Assay Details
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239960.2 | XP_006240022.1 | 1 - 2 | 229 | 75 | |
XM_006239962.2 | XP_006240024.1 | 1 - 2 | 228 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006239960.2, XM_006239962.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
