Assay Details
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239962.2 | XP_006240024.1 | 6 - 7 | 681 | 102 | |
GenBank mRNA | BC158664.1 | - | 478 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
