Assay Details
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100717.1 | NP_001094187.1 | 1 - 2 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
