Assay Details
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239913.2 | XP_006239975.1 | 2 - 3 | 151 | 115 | |
GenBank mRNA | AY211264.1 | - | 2 - 3 | 90 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY211264.1, XM_006239913.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
