Assay Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239786.1 | XP_006239848.1 | 14 - 15 | 1703 | 96 | |
XM_006239788.1 | XP_006239850.1 | 14 - 15 | 1359 | 96 | ||
XM_006239789.2 | XP_006239851.1 | 14 - 15 | 1634 | 96 | ||
GenBank mRNA | BC060545.1 | - | 14 - 15 | 1286 | 96 | |
D16479.1 | - | 13 - 14 | 1250 | 96 | ||
FM105991.1 | - | 5 - 6 | 430 | 96 | ||
FQ226284.1 | - | 2 - 3 | 137 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
