Assay Details
Gene Symbol: Sdsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108336.1 | NP_001101806.1 | 7 - 8 | 1057 | 105 | |
XM_006249399.2 | XP_006249461.1 | 5 - 6 | 1038 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
