Assay Details
Gene Symbol: Styxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037788.1 | NP_001032877.1 | 2 - 3 | 130 | 68 | |
XM_006249180.2 | XP_006249242.1 | 2 - 3 | 350 | 68 | ||
XM_006249181.2 | XP_006249243.1 | 2 - 3 | 312 | 68 | ||
XM_008769143.1 | XP_008767365.1 | 2 - 3 | 309 | 68 | ||
XM_008769146.1 | XP_008767368.1 | 2 - 3 | 350 | 68 | ||
GenBank mRNA | BC098678.1 | - | 2 - 3 | 130 | 7382696 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008769143.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
