Assay Details
Gene Symbol: Styxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249180.2 | XP_006249242.1 | 1 - 2 | 250 | 96 | |
XM_008769146.1 | XP_008767368.1 | 1 - 2 | 250 | 96 | ||
XM_008769148.1 | XP_008767370.1 | 1 - 2 | 250 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006249180.2, XM_008769146.1, XM_008769148.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
