Assay Details
Gene Symbol: Styxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037788.1 | NP_001032877.1 | 5 - 6 | 502 | 91 | |
XM_006249180.2 | XP_006249242.1 | 5 - 6 | 725 | 91 | ||
XM_006249181.2 | XP_006249243.1 | 5 - 6 | 687 | 91 | ||
XM_008769143.1 | XP_008767365.1 | 6 - 7 | 880 | 91 | ||
XM_008769144.1 | XP_008767366.1 | 4 - 5 | 571 | 91 | ||
XM_008769146.1 | XP_008767368.1 | 5 - 6 | 725 | 91 | ||
XM_008769147.1 | XP_008767369.1 | 3 - 4 | 539 | 91 | ||
XM_008769148.1 | XP_008767370.1 | 3 - 4 | 497 | 91 | ||
GenBank mRNA | BC098678.1 | - | 5 - 6 | 502 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
