Assay Details
Gene Symbol: Pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025272.1 | NP_001020443.1 | 3 - 4 | 524 | 69 | |
XM_006249534.2 | XP_006249596.1 | 3 - 4 | 556 | 69 | ||
GenBank mRNA | BC092607.1 | - | 3 - 4 | 524 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
