Assay Details
Gene Symbol: Ccdc62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134766.1 | NP_001128238.1 | 1 - 2 | 169 | 75 | |
XM_006249355.2 | XP_006249417.1 | 1 - 2 | 1037 | 75 | ||
XM_006249356.2 | XP_006249418.1 | 1 - 2 | 1037 | 75 | ||
XM_006249357.2 | XP_006249419.1 | 1 - 2 | 1037 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
