Assay Details
Gene Symbol: Ccdc62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134766.1 | NP_001128238.1 | 2 - 3 | 359 | 82 | |
XM_006249355.2 | XP_006249417.1 | 2 - 3 | 1227 | 82 | ||
XM_006249356.2 | XP_006249418.1 | 2 - 3 | 1227 | 82 | ||
XM_006249357.2 | XP_006249419.1 | 2 - 3 | 1227 | 82 | ||
XM_006249358.1 | XP_006249420.1 | 2 - 3 | 300 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
