Assay Details
Gene Symbol: Cox6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012814.1 | NP_036946.1 | 1 - 2 | 129 | 133 | |
GenBank mRNA | CB716915.1 | - | 1 - 2 | 129 | 133 | |
FQ213920.1 | - | 1 - 2 | 116 | 133 | ||
FQ221354.1 | - | 1 - 2 | 116 | 133 | ||
FQ221968.1 | - | 1 - 2 | 118 | 133 | ||
FQ224366.1 | - | 1 - 2 | 116 | 133 | ||
FQ225833.1 | - | 1 - 2 | 116 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
