Assay Details
Gene Symbol: Prkab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249481.2 | XP_006249543.1 | 1 - 2 | 121 | 101 | |
GenBank mRNA | X95577.1 | - | 1 - 2 | 91 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the X95577.1, XM_006249481.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
