Assay Details
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012669.1 | NP_036801.1 | 7 - 8 | 1650 | 85 | |
GenBank mRNA | J03170.1 | - | 7 - 8 | 1650 | 85 | |
X53297.1 | - | 6 - 7 | 1008 | 85 | ||
X54423.1 | - | 7 - 8 | 1500 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
