Assay Details
Gene Symbol: Lrch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127551.1 | NP_001121023.1 | 14 - 15 | 1598 | 116 | |
XM_006249175.2 | XP_006249237.1 | 14 - 15 | 1614 | 116 | ||
GenBank mRNA | BC166781.1 | - | 14 - 15 | 1598 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
