Assay Details
Gene Symbol: Fkbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105922.1 | NP_001099392.1 | 2 - 3 | 206 | 68 | |
XM_006249131.2 | XP_006249193.1 | 2 - 3 | 225 | 68 | ||
XM_008769130.1 | XP_008767352.1 | 2 - 3 | 225 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
