Assay Details
Gene Symbol: Fkbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105922.1 | NP_001099392.1 | 8 - 9 | 1011 | 69 | |
XM_006249131.2 | XP_006249193.1 | 8 - 9 | 1030 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein peptidyl-prolyl isomerization
protein folding
spermatogenesis
peptidyl-proline modification
cell differentiation
gene silencing by RNA
piRNA metabolic process
DNA methylation involved in gamete generation
meiotic cell cycle
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
FK506 binding
Hsp90 protein binding
protein peptidyl-prolyl isomerization
protein folding
spermatogenesis
peptidyl-proline modification
cell differentiation
gene silencing by RNA
piRNA metabolic process
DNA methylation involved in gamete generation
meiotic cell cycle
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
FK506 binding
Hsp90 protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P126055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143778 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P126055 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P126055 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P126055 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P126055 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22944 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24630 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37992 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10757696 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757694 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757691 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757695 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757693 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757692 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757689 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757697 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10757688 | - | - |
Affymetrix Rat Clariom S Assay | PSR1200003806.rn.1 | TC1200000425.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10757688 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21421523 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649858 | - | - |
