Assay Details
Gene Symbol: Rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108333.1 | NP_001101803.1 | 5 - 6 | 968 | 93 | |
XM_006249238.2 | XP_006249300.1 | 6 - 7 | 1063 | 93 | ||
XM_006249239.2 | XP_006249301.1 | 6 - 7 | 815 | 93 | ||
XM_006249240.2 | XP_006249302.1 | 6 - 7 | 1052 | 93 | ||
XM_008769150.1 | XP_008767372.1 | 6 - 7 | 1059 | 93 | ||
GenBank mRNA | BC168697.1 | - | 3 - 4 | 367 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
