Assay Details
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006957.2 | NP_001006958.1 | 4 - 5 | 444 | 117 | |
GenBank mRNA | BC083818.1 | - | 4 - 5 | 436 | 7110979 | 117 |
CB718613.1 | - | 4 - 5 | 444 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
