Assay Details
Gene Symbol: Crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053670.3 | NP_446122.1 | 2 - 3 | 61 | 91 | |
GenBank mRNA | AF440799.2 | - | 2 - 3 | 61 | 91 | |
BC059117.1 | - | 2 - 3 | 165 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
