Assay Details
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013046.1 | NP_001013064.1 | 2 - 3 | 209 | 82 | |
XM_008769221.1 | XP_008767443.1 | 2 - 3 | 259 | 82 | ||
GenBank mRNA | BC085769.1 | - | 2 - 3 | 209 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
