Assay Details
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013046.1 | NP_001013064.1 | 5 - 6 | 586 | 77 | |
GenBank mRNA | BC085769.1 | - | 5 - 6 | 586 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
