Assay Details
Gene Symbol: Coq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039022.1 | NP_001034111.1 | 1 - 2 | 206 | 62 | |
GenBank mRNA | BC098719.1 | - | 1 - 2 | 206 | 7370143 | 62 |
FQ219423.1 | - | 1 - 2 | 211 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
