Assay Details
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_222147.8 | XP_222147.4 | 1 - 2 | 195 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_222147.8 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
