Assay Details
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769205.1 | XP_008767427.1 | 6 - 7 | 735 | 95 | |
XM_222147.8 | XP_222147.4 | 6 - 7 | 590 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
