Assay Details
Gene Symbol: Mios | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173554.1 | NP_001167025.1 | 6 - 7 | 2128 | 93 | |
XM_006236090.2 | XP_006236152.1 | 5 - 6 | 2128 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
