Assay Details
Gene Symbol: Tac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124768.1 | NP_001118240.1 | 4 - 5 | 346 | 99 | |
NM_012666.2 | NP_036798.1 | 5 - 6 | 391 | 144 | ||
XM_008762692.1 | XP_008760914.1 | 5 - 6 | 532 | 144 | ||
XM_008762693.1 | XP_008760915.1 | 4 - 5 | 484 | 99 | ||
GenBank mRNA | M14312.1 | - | 3 - 4 | 258 | 99 | |
M15191.1 | - | 5 - 6 | 393 | 144 | ||
M34183.1 | - | 4 - 5 | 348 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
