Assay Details
Gene Symbol: Adck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107855.1 | NP_001101325.1 | 2 - 3 | 1452 | 70 | |
XM_008762782.1 | XP_008761004.1 | 2 - 3 | 1475 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
