Assay Details
Gene Symbol: Adck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107855.1 | NP_001101325.1 | 7 - 8 | 2122 | 72 | |
XM_006236326.2 | XP_006236388.1 | 6 - 7 | 1998 | 72 | ||
GenBank mRNA | BC161918.1 | - | 6 - 7 | 1910 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
