Assay Details
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053787.2 | NP_446239.2 | 8 - 9 | 1264 | 89 | |
XM_006236376.2 | XP_006236438.1 | 6 - 7 | 840 | 89 | ||
GenBank mRNA | AB032019.1 | - | 8 - 9 | 1264 | 89 | |
AF209196.1 | - | 1215 | 89 | |||
AY762624.1 | - | 7 - 8 | 952 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
