Assay Details
Gene Symbol: Seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY325252.1 | - | 6485 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P283346 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21963 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21963 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P283346 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P283346 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21963 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P283346 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21963 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21963 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P283346 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87365 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29044 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19384 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10802486 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802498 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802483 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802496 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802502 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802500 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802503 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802505 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802497 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802501 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802494 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802493 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802504 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802485 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802499 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802491 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802495 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802492 | - | - |
Rat Genome 230 2.0 Array | 1385634_at | - | - |
Rat Genome 230 2.0 Array | 1398988_at | - | - |
Rat Genome 230 2.0 Array | 1385634_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398988_PM_at | - | - |
Rat Expression Set 230 | 1398988_at | - | - |
Rat Expression Set 230 | 1398988_PM_at | - | - |
Rat Expression Set 230 | 1385634_at | - | - |
Rat Expression Set 230 | 1385634_PM_at | - | - |
Rat Genome U34 Set | rc_AI058367_at | - | - |
Rat Genome U34 Set | rc_AA817959_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650160 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650327 | - | - |
