Assay Details
Gene Symbol: Napg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107384.1 | NP_001100854.1 | 1 - 2 | 165 | 93 | |
XM_006254829.2 | XP_006254891.1 | 1 - 2 | 162 | 93 | ||
GenBank mRNA | AY724541.1 | - | 1 - 2 | 67 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107384.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
