Assay Details
Gene Symbol: Napg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107384.1 | NP_001100854.1 | 2 - 3 | 236 | 78 | |
XM_006254829.2 | XP_006254891.1 | 2 - 3 | 233 | 78 | ||
GenBank mRNA | AY724541.1 | - | 2 - 3 | 138 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107384.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
