Assay Details
Gene Symbol: Napg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107384.1 | NP_001100854.1 | 6 - 7 | 541 | 133 | |
XM_006254829.2 | XP_006254891.1 | 7 - 8 | 533 | 133 | ||
GenBank mRNA | AY724541.1 | - | 7 - 8 | 438 | 133 | |
FQ211390.1 | - | 4 - 5 | 415 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
