Assay Details
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169120.1 | NP_001162591.1 | 8 - 9 | 1015 | 142 | |
XM_006231127.2 | XP_006231189.1 | 8 - 9 | 1269 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
