Assay Details
Gene Symbol: Psat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198738.2 | NP_942033.2 | 4 - 5 | 416 | 115 | |
GenBank mRNA | AY239016.1 | - | 4 - 5 | 404 | 115 | |
BC079352.1 | - | 4 - 5 | 416 | 7108218 | 115 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
