Assay Details
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024887.1 | NP_001020058.1 | 4 - 5 | 533 | 58 | |
XM_006230552.2 | XP_006230614.1 | 4 - 5 | 1367 | 58 | ||
XM_006230553.2 | XP_006230615.1 | 4 - 5 | 999 | 58 | ||
XM_008759988.1 | XP_008758210.1 | 3 - 4 | 2565 | 58 | ||
XM_008759989.1 | XP_008758211.1 | 4 - 5 | 1058 | 58 | ||
XM_008759990.1 | XP_008758212.1 | 4 - 5 | 561 | 58 | ||
GenBank mRNA | BC097488.1 | - | 4 - 5 | 533 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
