Assay Details
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011941.1 | NP_001011941.1 | 3 - 4 | 606 | 176 | |
XM_006231201.2 | XP_006231263.1 | 4 - 5 | 765 | 176 | ||
XM_006231202.2 | XP_006231264.1 | 4 - 5 | 765 | 176 | ||
XM_008760322.1 | XP_008758544.1 | 4 - 5 | 765 | 176 | ||
GenBank mRNA | BC083761.1 | - | 3 - 4 | 606 | 176 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
