Assay Details
Gene Symbol: Ccdc88b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_215206.9 | XP_215206.6 | 22 - 23 | 3894 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
