Assay Details
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108516.1 | NP_001101986.1 | 1 - 2 | 87 | 91 | |
XM_006230850.1 | XP_006230912.1 | 1 - 2 | 112 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108516.1, XM_006230850.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
