Assay Details
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108516.1 | NP_001101986.1 | 2 - 3 | 251 | 87 | |
XM_006230850.1 | XP_006230912.1 | 2 - 3 | 276 | 87 | ||
GenBank mRNA | BC166788.1 | - | 2 - 3 | 198 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
