Assay Details
Gene Symbol: Rce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100585.1 | NP_001094055.1 | 2 - 3 | 302 | 87 | |
XM_006230788.2 | XP_006230850.1 | 2 - 3 | 332 | 87 | ||
XM_006230790.2 | XP_006230852.1 | 2 - 3 | 332 | 87 | ||
XM_006230791.2 | XP_006230853.1 | 2 - 3 | 332 | 87 | ||
XM_008760135.1 | XP_008758357.1 | 2 - 3 | 332 | 87 | ||
GenBank mRNA | BC087629.1 | - | 2 - 3 | 289 | 87 | |
BC158593.1 | - | 2 - 3 | 302 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
