Assay Details
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108505.2 | NP_001101975.2 | 2 - 3 | 264 | 128 | |
GenBank mRNA | FQ227185.1 | - | 2 - 3 | 255 | 128 | |
FQ228402.1 | - | 2 - 3 | 262 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
