Assay Details
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108505.2 | NP_001101975.2 | 3 - 4 | 341 | 74 | |
GenBank mRNA | FQ227185.1 | - | 3 - 4 | 332 | 74 | |
FQ228402.1 | - | 3 - 4 | 339 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
