Assay Details
Gene Symbol: Glrx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032614.2 | NP_116003.2 | 9 - 10 | 883 | 156 | |
XM_006230497.2 | XP_006230559.1 | 9 - 10 | 1296 | 156 | ||
GenBank mRNA | AF118651.1 | - | 9 - 10 | 717 | 156 | |
BC086381.1 | - | 9 - 10 | 883 | 156 | ||
FQ216152.1 | - | 6 - 7 | 533 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
