Assay Details
Gene Symbol: Glrx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032614.2 | NP_116003.2 | 8 - 9 | 836 | 96 | |
XM_006230497.2 | XP_006230559.1 | 8 - 9 | 1249 | 96 | ||
GenBank mRNA | AF118651.1 | - | 8 - 9 | 670 | 96 | |
BC086381.1 | - | 8 - 9 | 836 | 96 | ||
FQ216152.1 | - | 5 - 6 | 486 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
