Assay Details
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034134.1 | NP_001029306.1 | 1 - 2 | 324 | 58 | |
GenBank mRNA | BC100264.1 | - | 1 - 2 | 324 | 58 | |
FQ209474.1 | - | 349 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
