Assay Details
Gene Symbol: Pdgfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031524.1 | NP_113712.1 | 2 - 3 | 548 | 88 | |
GenBank mRNA | CB580369.1 | - | 2 - 3 | 558 | 88 | |
Z14117.1 | - | 2 - 3 | 129 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
