Assay Details
Gene Symbol: Pdgfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031524.1 | NP_113712.1 | 5 - 6 | 992 | 70 | |
GenBank mRNA | Z14117.1 | - | 5 - 6 | 573 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
embryonic placenta development
positive regulation of endothelial cell proliferation
monocyte chemotaxis
positive regulation of glomerular filtration
DNA replication
protein phosphorylation
substrate-dependent cell migration
heart development
blood coagulation
positive regulation of cell proliferation
response to wounding
response to organic substance
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
positive regulation of phosphatidylinositol 3-kinase signaling
response to organic cyclic compound
positive regulation of smooth muscle cell migration
cell growth
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
cell projection assembly
actin cytoskeleton organization
hemopoiesis
positive regulation of cell migration
negative regulation of cell migration
positive regulation of protein autophosphorylation
activation of protein kinase activity
activation of protein kinase B activity
response to estradiol
response to insulin
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
paracrine signaling
wound healing
regulation of cell proliferation
response to drug
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of blood vessel endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase activity
response to estrogen
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of DNA replication
positive regulation of fibroblast growth factor receptor signaling pathway
positive regulation of mitotic nuclear division
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle, embryonic
platelet-derived growth factor receptor signaling pathway
positive regulation of fibroblast proliferation
blood vessel morphogenesis
positive regulation of smooth muscle cell proliferation
regulation of peptidyl-tyrosine phosphorylation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of chemotaxis
cell chemotaxis
branching involved in salivary gland morphogenesis
epithelial cell proliferation involved in salivary gland morphogenesis
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
cellular response to growth factor stimulus
cellular response to mycophenolic acid
positive regulation of glomerular mesangial cell proliferation
metanephric glomerular mesangial cell development
metanephric glomerular mesangial cell proliferation involved in metanephros development
metanephric glomerular endothelium development
reactive oxygen species metabolic process
positive regulation of calcium ion import
positive regulation of hyaluronan biosynthetic process
positive regulation of reactive oxygen species metabolic process
positive regulation of DNA biosynthetic process
positive regulation of metanephric mesenchymal cell migration
receptor binding
platelet-derived growth factor receptor binding
collagen binding
growth factor activity
superoxide-generating NADPH oxidase activator activity
chemoattractant activity
identical protein binding
protein homodimerization activity
protein heterodimerization activity
platelet-derived growth factor binding
response to hypoxia
embryonic placenta development
positive regulation of endothelial cell proliferation
monocyte chemotaxis
positive regulation of glomerular filtration
DNA replication
protein phosphorylation
substrate-dependent cell migration
heart development
blood coagulation
positive regulation of cell proliferation
response to wounding
response to organic substance
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
positive regulation of phosphatidylinositol 3-kinase signaling
response to organic cyclic compound
positive regulation of smooth muscle cell migration
cell growth
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
cell projection assembly
actin cytoskeleton organization
hemopoiesis
positive regulation of cell migration
negative regulation of cell migration
positive regulation of protein autophosphorylation
activation of protein kinase activity
activation of protein kinase B activity
response to estradiol
response to insulin
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
paracrine signaling
wound healing
regulation of cell proliferation
response to drug
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of blood vessel endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase activity
response to estrogen
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of DNA replication
positive regulation of fibroblast growth factor receptor signaling pathway
positive regulation of mitotic nuclear division
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle, embryonic
platelet-derived growth factor receptor signaling pathway
positive regulation of fibroblast proliferation
blood vessel morphogenesis
positive regulation of smooth muscle cell proliferation
regulation of peptidyl-tyrosine phosphorylation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of chemotaxis
cell chemotaxis
branching involved in salivary gland morphogenesis
epithelial cell proliferation involved in salivary gland morphogenesis
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
cellular response to growth factor stimulus
cellular response to mycophenolic acid
positive regulation of glomerular mesangial cell proliferation
metanephric glomerular mesangial cell development
metanephric glomerular mesangial cell proliferation involved in metanephros development
metanephric glomerular endothelium development
reactive oxygen species metabolic process
positive regulation of calcium ion import
positive regulation of hyaluronan biosynthetic process
positive regulation of reactive oxygen species metabolic process
positive regulation of DNA biosynthetic process
positive regulation of metanephric mesenchymal cell migration
receptor binding
platelet-derived growth factor receptor binding
collagen binding
growth factor activity
superoxide-generating NADPH oxidase activator activity
chemoattractant activity
identical protein binding
protein homodimerization activity
protein heterodimerization activity
platelet-derived growth factor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P086538 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P080479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31344 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P086538 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P080479 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P086538 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P080479 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P080479 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P086538 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P223549 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50844 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10905522 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905525 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905524 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905523 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905526 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10905521 | - | - |
Affymetrix Rat Clariom S Assay | PSR0700027761.rn.1 | TC0700003519.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700027762.rn.1 | TC0700003519.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10905521 | - | - |
Rat Genome U34 Set | L41623cds_s_at | - | - |
Rat Genome U34 Set | Z14117cds_i_at | - | - |
Rat Genome U34 Set | L41623cds_s_at | - | - |
Rat Genome U34 Set | Z14117cds_i_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22342637 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357500 | - | - |
